CH6371381551 Chem Impex International, Inc.

(S)-3-Amino-3-(3-nitrophenyl)propionic acid

  • MDL number MFCD04113693
  • Cas Number : 734529-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-beta-Phe(3-NO2)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-3-nitrophenyl-propionic-acid/6978?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-3-nitrophenyl-propionic-acid/6978?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart