CH6371381553 Chem Impex International, Inc.

(R)-3-Amino-3-(2-methylphenyl)propionic acid

  • MDL number MFCD04113646
  • Cas Number : 752198-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-beta-Phe(2-Me)-OH
Purity 0.98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-2-methylphenyl-propionic-acid/6987?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-2-methylphenyl-propionic-acid/6987?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart