CH6371381559 Chem Impex International, Inc.

(R)-3-Amino-3-(4-methylphenyl)propionic acid

  • MDL number MFCD03840389
  • Cas Number : 479064-87-4
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-240 °C (dec.)
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-4-methylphenyl-propionic-acid/6999?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-4-methylphenyl-propionic-acid/6999?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart