CH6371381561 Chem Impex International, Inc.

Boc-(R)-3-amino-3-(2-methoxyphenyl)propionic acid

  • MDL number MFCD03427936
  • Cas Number : 500788-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-beta-Phe(2-OMe)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-2-methoxyphenyl-propionic-acid/7003?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-2-methoxyphenyl-propionic-acid/7003?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart