CH6371381565 Chem Impex International, Inc.

(R)-3-Amino-3-(3-methoxyphenyl)propionic acid

  • MDL number MFCD04113658
  • Cas Number : 765895-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-3-methoxyphenyl-propionic-acid/7007?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-3-methoxyphenyl-propionic-acid/7007?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart