CH6371381571 Chem Impex International, Inc.

Fmoc-(S)-3-amino-3-(4-methoxyphenyl)propionic acid

  • MDL number MFCD03427976
  • Cas Number : 501015-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-3-4-methoxyphenyl-propionic-acid/7016?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-3-4-methoxyphenyl-propionic-acid/7016?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart