CH6371381573 Chem Impex International, Inc.

Boc-(R)-3-amino-3-(2-hydroxyphenyl)propionic acid

  • MDL number MFCD03427939
  • Cas Number : 500788-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-165 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-2-hydroxyphenyl-propionic-acid/7019?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-2-hydroxyphenyl-propionic-acid/7019?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart