CH6371381574 Chem Impex International, Inc.

Fmoc-(R)-3-amino-3-(2-hydroxyphenyl)propionic acid

  • MDL number MFCD03428015
  • Cas Number : 511272-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-164 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-3-2-hydroxyphenyl-propionic-acid/7021?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-3-2-hydroxyphenyl-propionic-acid/7021?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart