CH6371381578 Chem Impex International, Inc.

Fmoc-(S)-3-amino-3-(4-hydroxyphenyl)propionic acid

  • MDL number MFCD03427979
  • Cas Number : 501015-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 171 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-s-3-amino-3-4-hydroxyphenyl-propionic-acid/7031?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-s-3-amino-3-4-hydroxyphenyl-propionic-acid/7031?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart