CH6371381579 Chem Impex International, Inc.

(R)-3-Amino-3-(2,4-dichlorophenyl)propionic acid

  • MDL number MFCD04113640
  • Cas Number : 778571-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239-243 �dec.)
Synonym D-beta-Phe(2,4-DiCl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-2-4-dichlorophenyl-propionic-acid/7032?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-2-4-dichlorophenyl-propionic-acid/7032?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart