CH6371381580 Chem Impex International, Inc.

(S)-3-Amino-3-(2,4-dichlorophenyl)propionic acid

  • MDL number MFCD04113673
  • Cas Number : 757937-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-beta-Phe(2,4-DiCl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-2-4-dichlorophenyl-propionic-acid/7033?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-2-4-dichlorophenyl-propionic-acid/7033?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart