CH6371381581 Chem Impex International, Inc.

(S)-3-Amino-3-(2,3-dichlorophenyl)propionic acid

  • MDL number MFCD04113671
  • Cas Number : 748128-13-4
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white Solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-2-3-dichlorophenyl-propionic-acid/7039?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-2-3-dichlorophenyl-propionic-acid/7039?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart