CH6371381584 Chem Impex International, Inc.

(S)-3-Amino-3-(2,3-dimethoxyphenyl)propionic acid

  • MDL number MFCD04113672
  • Cas Number : 752198-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-201 �dec.)
Synonym L-beta-Phe(2,3-dimethoxy)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-2-3-dimethoxyphenyl-propionic-acid/7045?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-2-3-dimethoxyphenyl-propionic-acid/7045?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart