CH6371381586 Chem Impex International, Inc.

(R)-3-Amino-3-(3,5-dimethoxyphenyl)propionic acid

  • MDL number MFCD04113652
  • Cas Number : 744193-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199-203
Synonym D-beta-Phe(3,5-dimethoxy)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-3-5-dimethoxyphenyl-propionic-acid/7056?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-3-5-dimethoxyphenyl-propionic-acid/7056?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart