CH6371381587 Chem Impex International, Inc.

Fmoc-(S)-3-amino-3-(3-pyridyl)propionic acid

  • MDL number MFCD03427986
  • Cas Number : 507472-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-188
Synonym Fmoc-L-beta-Ala-(3-pyridyl)-OH
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-3-3-pyridyl-propionic-acid/7064?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-3-3-pyridyl-propionic-acid/7064?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart