CH6371381590 Chem Impex International, Inc.

Fmoc-(R)-3-amino-3-(2-thienyl)propionic acid

  • MDL number MFCD03428026
  • Cas Number : 511272-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-187 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-3-2-thienyl-propionic-acid/7067?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-3-2-thienyl-propionic-acid/7067?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart