CH6371381592 Chem Impex International, Inc.

Boc-(R)-3-amino-3-(3-thienyl)propionic acid

  • MDL number MFCD03427951
  • Cas Number : 500788-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129 - 131 °C
Purity 97
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-3-thienyl-propionic-acid/7071?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-3-thienyl-propionic-acid/7071?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart