CH6371381597 Chem Impex International, Inc.

Fmoc-(R)-3-amino-3-(2-naphthyl)propionic acid

  • MDL number MFCD03428029
  • Cas Number : 511272-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-193 �C
Synonym Fmoc-D-beta-Ala-(2-naphthyl)-OH
Purity 99.5
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-3-2-naphthyl-propionic-acid/7084?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-3-2-naphthyl-propionic-acid/7084?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart