CH6371381598 Chem Impex International, Inc.

Fmoc-(S)-3-amino-3-(2-naphthyl)propionic acid

  • MDL number MFCD03427991
  • Cas Number : 507472-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-194 �C
Synonym Fmoc-(S)-3-amino-3-(2-naphthyl)propionic acid
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-3-2-naphthyl-propionic-acid/7085?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-3-2-naphthyl-propionic-acid/7085?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart