CH6371381599 Chem Impex International, Inc.

Boc-(R)-3-amino-3-(2-furyl)propionic acid

  • MDL number MFCD04117839
  • Cas Number : 1217725-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-75
Synonym Boc-L-beta-Ala-(2-furyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-2-furyl-propionic-acid/7087?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-3-amino-3-2-furyl-propionic-acid/7087?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart