CH6371381603 Chem Impex International, Inc.

Boc-D-2-amino-5-phenylpentanoic acid dicyclohexylamine salt

  • MDL number MFCD04117837

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-2-amino-5-phenylpentanoic-acid-dicyclohexylamine-salt/7092?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-2-amino-5-phenylpentanoic-acid-dicyclohexylamine-salt/7092?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart