CH6371381612 Chem Impex International, Inc.

D-2-Amino-5-phenylpentanoic acid

  • MDL number MFCD03844654
  • Cas Number : 2046-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-2-amino-5-phenylpentanoic-acid/7103?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-2-amino-5-phenylpentanoic-acid/7103?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart