CH6371381615 Chem Impex International, Inc.

(R)-3-Amino-3-(6-methoxy-3-pyridyl)propionic acid

  • MDL number MFCD04118072
  • Cas Number : 712321-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 200 °C (dec.)
Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-6-methoxy-3-pyridyl-propionic-acid/7106?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-amino-3-6-methoxy-3-pyridyl-propionic-acid/7106?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart