CH6371381616 Chem Impex International, Inc.

(S)-3-Amino-3-(6-methoxy-3-pyridyl)propionic acid

  • MDL number MFCD04118071
  • Cas Number : 877119-70-5
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-218 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-6-methoxy-3-pyridyl-propionic-acid/7107?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-3-6-methoxy-3-pyridyl-propionic-acid/7107?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart