CH6371381619 Chem Impex International, Inc.

Boc-(R)-a-(2-fluorobenzyl)proline

  • MDL number MFCD06795524
  • Cas Number : 959576-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-156
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-a-2-fluorobenzyl-proline/7113?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-a-2-fluorobenzyl-proline/7113?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart