CH6371381620 Chem Impex International, Inc.

Boc-(S)-alpha-(2-fluorobenzyl)proline

  • MDL number MFCD06659155
  • Cas Number : 1217635-13-6
  • Molecular Weight : 323.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-160 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-a-2-fluorobenzyl-proline/7115?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-a-2-fluorobenzyl-proline/7115?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart