CH6371381622 Chem Impex International, Inc.

Boc-(S)-a-(4-chlorobenzyl)proline

  • MDL number MFCD06796760
  • Cas Number : 1217777-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-alpha-4-chlorobenzyl-proline/7123?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-alpha-4-chlorobenzyl-proline/7123?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart