CH6371381627 Chem Impex International, Inc.

Boc-(S)-(2-nitrobenzyl)proline

  • MDL number MFCD07363495
  • Cas Number : 1217814-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-136 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-2-nitrobenzyl-proline/7141?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-2-nitrobenzyl-proline/7141?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart