CH6371381629 Chem Impex International, Inc.

(S)-a-(4-Trifluoromethylbenzyl)proline�

  • MDL number MFCD06796803
  • Cas Number : 1217861-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-alpha-4-trifluoromethylbenzyl-proline-font-size-2-face-symbol-middot-font-hcl/7173?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-alpha-4-trifluoromethylbenzyl-proline-font-size-2-face-symbol-middot-font-hcl/7173?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart