CH6371381631 Chem Impex International, Inc.

Boc-(S)-a-(4-biphenylmethyl)proline

  • MDL number MFCD06659364
  • Cas Number : 1217857-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-164
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-alpha-4-biphenylmethyl-proline/7210?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-alpha-4-biphenylmethyl-proline/7210?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart