CH6371381632 Chem Impex International, Inc.

(S)-alpha-(3-Pyridinylmethyl)proline 2HCl

  • MDL number MFCD06659377
  • Cas Number : 1217830-70-0
  • Molecular Weight : 279.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-a-3-pyridinylmethyl-proline·2hcl/7240?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-a-3-pyridinylmethyl-proline·2hcl/7240?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart