CH6371381637 Chem Impex International, Inc.

(R)-a-Propynylproline�

  • MDL number MFCD06659239
  • Cas Number : 1049733-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-a-propynylproline-middot-hcl/7250?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-a-propynylproline-middot-hcl/7250?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart