CH6371381638 Chem Impex International, Inc.

(R)-gamma-Benzyl-L-proline HCl

  • MDL number MFCD06659385
  • Cas Number : 1279049-67-0
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-g-benzyl-l-proline-middot-hcl/7254?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-g-benzyl-l-proline-middot-hcl/7254?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart