CH6371381643 Chem Impex International, Inc.

(R)-gamma-(4-Chlorobenzyl)-L-proline HCl

  • MDL number MFCD06659396
  • Cas Number : 1049733-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-gamma-4-chlorobenzyl-l-proline-font-size-2-face-symbol-middot-font-hcl/7266?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-gamma-4-chlorobenzyl-l-proline-font-size-2-face-symbol-middot-font-hcl/7266?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart