CH6371381647 Chem Impex International, Inc.

(R)-gamma-(4-Methylbenzyl)-L-proline HCl

  • MDL number MFCD06659408
  • Cas Number : 1049734-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-236 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-gamma-4-methylbenzyl-l-proline-font-size-2-face-symbol-middot-font-hcl/7278?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-gamma-4-methylbenzyl-l-proline-font-size-2-face-symbol-middot-font-hcl/7278?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart