CH6371381648 Chem Impex International, Inc.

(R)-gamma-(3-Nitrobenzyl)-L-proline HCl

  • MDL number MFCD06659412
  • Cas Number : 1049740-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-164 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-gamma-3-nitrobenzyl-l-proline-font-size-2-face-symbol-middot-font-hcl/7282?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-gamma-3-nitrobenzyl-l-proline-font-size-2-face-symbol-middot-font-hcl/7282?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart