CH6371381654 Chem Impex International, Inc.

Boc-(R)-gamma-(4-cyanobenzyl)-L-proline

  • MDL number MFCD06796826
  • Cas Number : 1217684-11-1
  • Molecular Weight : 330.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-r-i-gamma-4-cyanobenzyl-l-proline/7307?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-r-i-gamma-4-cyanobenzyl-l-proline/7307?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart