CH6371381661 Chem Impex International, Inc.

(�rans-4-(2-Fluorophenyl)pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD06659255
  • Cas Number : 1820569-65-0
  • Molecular Weight : 245.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-194 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/plusmn-i-trans-i-4-2-fluorophenyl-pyrrolidine-3-carboxylic-acid-hydrochloride/7345?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/plusmn-i-trans-i-4-2-fluorophenyl-pyrrolidine-3-carboxylic-acid-hydrochloride/7345?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart