CH6371381670 Chem Impex International, Inc.

Boc-(�rans-4-(4-bromophenyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD07363505
  • Cas Number : 1217829-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-trans-DL-beta-Pro-4-(4-bromophenyl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-plusmn-i-trans-i-4-4-bromophenyl-pyrrolidine-3-carboxylic-acid/7361?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-plusmn-i-trans-i-4-4-bromophenyl-pyrrolidine-3-carboxylic-acid/7361?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart