CH6371381681 Chem Impex International, Inc.

Boc-(�rans-4-(2,5-dichlorophenyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD06659285
  • Cas Number : 1820572-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-180 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-plusmn-i-trans-i-4-2-5-dichlorophenyl-pyrrolidine-3-carboxylic-acid/7379?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-plusmn-i-trans-i-4-2-5-dichlorophenyl-pyrrolidine-3-carboxylic-acid/7379?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart