CH6371381699 Chem Impex International, Inc.

Boc-(�rans-4-(6-methoxy-3-pyridinyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD06659321
  • Cas Number : 1392213-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 130 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-plusmn-i-trans-i-4-6-methoxy-3-pyridinyl-pyrrolidine-3-carboxylic-acid/7415?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-plusmn-i-trans-i-4-6-methoxy-3-pyridinyl-pyrrolidine-3-carboxylic-acid/7415?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart