CH6371381704 Chem Impex International, Inc.

(�rans-4-(3-Thienyl)pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD06659325
  • Cas Number : 1864003-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/plusmn-i-trans-i-4-3-thienyl-pyrrolidine-3-carboxylic-acid-hydrochloride/7420?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/plusmn-i-trans-i-4-3-thienyl-pyrrolidine-3-carboxylic-acid-hydrochloride/7420?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart