CH6371381710 Chem Impex International, Inc.

2-(4-Boc-piperazinyl)-2-(3-fluorophenyl)acetic acid

  • MDL number MFCD06659334
  • Cas Number : 885272-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-170
Synonym alpha-(4-Boc-piperazinyl)-alpha-(3-fluorophenyl)acetic acid
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-boc-piperazinyl-2-3-fluorophenyl-acetic-acid/7426?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-boc-piperazinyl-2-3-fluorophenyl-acetic-acid/7426?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart