CH6371381712 Chem Impex International, Inc.

2-(4-Boc-piperazinyl)-2-(3-chlorophenyl)acetic acid

  • MDL number MFCD06659336
  • Cas Number : 885272-99-1
  • Molecular Weight : 354.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 142 °C
Purity 97
Storage temp. Store at 0 - 8 °C
Physical Form Yellowish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-boc-piperazinyl-2-3-chlorophenyl-acetic-acid/7429?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-boc-piperazinyl-2-3-chlorophenyl-acetic-acid/7429?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart