CH6371381727 Chem Impex International, Inc.

1-(Boc-amino)-4,9-dioxa-12-dodecanamine

  • MDL number MFCD04973131
  • Cas Number : 275823-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97100
Storage temp. Store at 0-8 °C
Physical Form Light yellow oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-boc-amino-4-9-dioxa-12-dodecanamine/7465?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-boc-amino-4-9-dioxa-12-dodecanamine/7465?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart