CH6371381730 Chem Impex International, Inc.

1-(Fmoc-amino)-4-methyl-cyclohex-3-ene-1-carboxylic acid

  • MDL number MFCD04973172
  • Cas Number : 1263047-06-8
  • Molecular Weight : 377.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-167 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-fmoc-amino-4-methyl-cyclohex-3-ene-1-carboxylic-acid/7468?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-fmoc-amino-4-methyl-cyclohex-3-ene-1-carboxylic-acid/7468?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart