CH6371381731 Chem Impex International, Inc.

1-Boc-2-triphenylphosphonium-hydrazine bromide

  • MDL number MFCD04973132
  • Cas Number : 356534-53-7
  • Molecular Weight : 473.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-188 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-boc-2-triphenylphosphonium-hydrazine-bromide/7469?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-boc-2-triphenylphosphonium-hydrazine-bromide/7469?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart