CH6371381735 Chem Impex International, Inc.

2-(tert-Butoxy)ethylamine HCl

  • MDL number MFCD04973191
  • Cas Number : 335598-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-117 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-tert-butoxy-ethylamine-font-size-2-face-symbol-middot-font-hcl/7473?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-tert-butoxy-ethylamine-font-size-2-face-symbol-middot-font-hcl/7473?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart