CH6371381740 Chem Impex International, Inc.

Nalpha-Allyloxycarbonyl-Nepsilon-Boc-D-lysine dicyclohexylammonium salt

  • MDL number MFCD04973096
  • Cas Number : 1423017-84-8
  • Molecular Weight : 511.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-147 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-allyloxycarbonyl-i-n-i-sup-epsilon-sup-boc-d-lysine-dicyclohexylammonium-salt/7479?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-allyloxycarbonyl-i-n-i-sup-epsilon-sup-boc-d-lysine-dicyclohexylammonium-salt/7479?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart