CH6371381741 Chem Impex International, Inc.

Nalpha-Allyloxycarbonyl-6-hydroxy-L-norleucine dicyclohexylammonium salt

  • MDL number MFCD06796878
  • Cas Number : 1263045-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-110 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-allyloxycarbonyl-6-hydroxy-l-norleucine-dicyclohexylammonium-salt/7480?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-allyloxycarbonyl-6-hydroxy-l-norleucine-dicyclohexylammonium-salt/7480?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart